Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan11
Entrez Gene ID
564029
Gene Name
tetraspanin 11
Gene Aliases
si:dkey-21h14.7
Chromosome Location
Chr.4: 11173014 - 11198355 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045037.1 NP_001038502.1 3 - 4 344 75
XM_005164652.4 XP_005164709.1 3 - 4 413 75
XM_005164653.4 XP_005164710.1 3 - 4 430 75

View Full Product Details