Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gli2a
Entrez Gene ID
30154
Gene Name
GLI family zinc finger 2a
Gene Aliases
fc85b05, gli2, wu:fc85b05, yot, you-too
Chromosome Location
Chr.9: 37141836 - 37296908 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130967.1 NP_571042.1 2 - 3 476 79
GenBank mRNA AF085746.2 - 2 - 3 476 79
BC163542.1 - 2 - 3 467 79

View Full Product Details