Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsp90ab1
Entrez Gene ID
30573
Gene Name
heat shock protein 90, alpha (cytosolic), class B member 1
Gene Aliases
hsp90b, hsp90beta, wu:fa29f01, wu:fa91e11, wu:fd59e11, wu:gcd22h07
Chromosome Location
Chr.20: 51198445 - 51211840 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131310.3 NP_571385.2 2 - 3 267 146
GenBank mRNA AF042108.1 - 2 - 3 187 146
AF068772.1 - 2 - 3 206 146
BC065359.1 - 2 - 3 213 146
BC090756.1 - 2 - 3 956 146
EH451195.1 - 2 - 3 299 146

View Full Product Details