Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mhc1uba
Entrez Gene ID
30757
Gene Name
major histocompatibility complex class I UBA
Gene Aliases
zgc:91955
Chromosome Location
Chr.19: 7072854 - 7101100 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131471.1 NP_571546.1 4 - 5 955 104
GenBank mRNA BC074095.1 - 4 - 5 929 104
BC164159.1 - 4 - 5 916 104
Z46777.1 - - 955 104

View Full Product Details