Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ociad1
Entrez Gene ID
393272
Gene Name
OCIA domain containing 1
Gene Aliases
zgc:56639, zgc:76992
Chromosome Location
Chr.20: 23245620 - 23253570 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200302.2 NP_956596.1 3 - 4 286 64
GenBank mRNA BC049514.1 - 3 - 4 288 64
BC066633.1 - 3 - 4 322 64

View Full Product Details