Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
efna1b
Entrez Gene ID
494151
Gene Name
ephrin-A1b
Gene Aliases
efna1
Chromosome Location
Chr.16: 23282660 - 23298149 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200783.3 NP_957077.2 3 - 4 639 94
GenBank mRNA AJ006838.1 - - 502 94
EH439178.1 - 3 - 4 665 94

View Full Product Details