Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdss2
Entrez Gene ID
436624
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
zgc:92156
Chromosome Location
Chr.13: 32660549 - 32726178 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002351.1 NP_001002351.1 6 - 7 1235 88
GenBank mRNA BC075908.1 - 6 - 7 1235 88

View Full Product Details