Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ism1
Entrez Gene ID
497617
Gene Name
isthmin 1
Gene Aliases
-
Chromosome Location
Chr.13: 34765169 - 34793487 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012376.1 NP_001012376.1 3 - 4 1338 58
GenBank mRNA AY856371.1 - 3 - 4 628 58
AY884208.1 - 3 - 4 1338 58
BC129141.1 - 3 - 4 1295 58

View Full Product Details