Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kif7
Entrez Gene ID
544651
Gene Name
kinesin family member 7
Gene Aliases
cos2, costal2
Chromosome Location
Chr.7: 14394515 - 14446588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014816.1 NP_001014816.1 10 - 11 2425 138
GenBank mRNA AY954727.1 - 10 - 11 2425 138

View Full Product Details