Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kif7
Entrez Gene ID
544651
Gene Name
kinesin family member 7
Gene Aliases
cos2, costal2
Chromosome Location
Chr.7: 14394515 - 14446588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014816.1 NP_001014816.1 12 - 13 2745 66
XM_005166379.4 XP_005166436.1 13 - 14 2916 66
GenBank mRNA AY954727.1 - 12 - 13 2745 66

Gene Symbol
LOC103909932
Entrez Gene ID
103909932
Gene Name
kinesin-like protein kif7
Gene Aliases
-
Chromosome Location
Chr.Unknown: 3650 - 13978 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009298287.3 XP_009296562.2 1 - 2 155 66

View Full Product Details