Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan2a
Entrez Gene ID
378854
Gene Name
tetraspanin 2a
Gene Aliases
sb:cb834, tspan2, wu:fb82d08, wu:fj10e11, zgc:110586
Chromosome Location
Chr.6: 206451 - 243323 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018150.1 NP_001018160.1 6 - 7 629 115
GenBank mRNA BC095323.1 - 6 - 7 629 115

View Full Product Details