Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scdb
Entrez Gene ID
553734
Gene Name
stearoyl-CoA desaturase b
Gene Aliases
zgc:112951
Chromosome Location
Chr.13: 30009617 - 30027708 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020705.1 NP_001018541.1 1 - 2 202 97
XM_005156845.4 XP_005156902.1 2 - 3 431 97
GenBank mRNA BC095851.1 - 1 - 2 202 97

View Full Product Details