Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnpy1
Entrez Gene ID
402819
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
-
Chromosome Location
Chr.2: 29986262 - 29996045 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039497.2 NP_001034586.1 3 - 4 508 148
GenBank mRNA AB201383.1 - 3 - 4 388 148
BC078218.1 - 3 - 4 487 148
BC091456.1 - 3 - 4 369 148
BC114321.1 - 3 - 4 458 148
BC122425.1 - 3 - 4 508 148
BC164976.1 - 2 - 3 255 148

View Full Product Details