Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnpy1
Entrez Gene ID
402819
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
-
Chromosome Location
Chr.2: 29986262 - 29996045 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039497.2 NP_001034586.1 4 - 5 714 103
GenBank mRNA AB055671.1 - 1 - 2 128 103
AB201383.1 - 4 - 5 594 103
BC078218.1 - 4 - 5 693 103
BC114321.1 - 4 - 5 664 103
BC122425.1 - 4 - 5 714 103
BC164976.1 - 3 - 4 461 103

View Full Product Details