Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glcci1a
Entrez Gene ID
378997
Gene Name
glucocorticoid induced 1a
Gene Aliases
glcci1, sb:cb902, testhymin, zgc:158237
Chromosome Location
Chr.19: 25352289 - 25427256 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079950.1 NP_001073419.1 2 - 3 886 61
GenBank mRNA AY835385.1 - 2 - 3 307 61
BC129170.1 - 2 - 3 886 61

View Full Product Details