Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elavl1b
Entrez Gene ID
30065
Gene Name
ELAV like RNA binding protein 1b
Gene Aliases
fb72d01, fk65g02, hug, wu:fb72d01, wu:fk65g02
Chromosome Location
Chr.11: 14955481 - 15001766 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308614.1 NP_001295543.1 7 - 8 886 66
NM_001308615.1 NP_001295544.1 6 - 7 826 66
NM_001308616.1 NP_001295545.1 6 - 7 813 66
NM_001308617.1 NP_001295546.1 7 - 8 876 66
NM_001308618.1 NP_001295547.1 5 - 6 722 66
NM_001308619.1 NP_001295548.1 6 - 7 785 66
NM_130909.2 NP_570984.2 7 - 8 889 66
XM_017358196.2 XP_017213685.1 6 - 7 887 66
XM_017358197.2 XP_017213686.1 5 - 6 735 66
GenBank mRNA AF184245.1 - 7 - 8 888 66
BC045277.1 - 5 - 6 659 66
BC165930.1 - 2 - 3 343 66

View Full Product Details