Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fabp3
Entrez Gene ID
171478
Gene Name
fatty acid binding protein 3, muscle and heart
Gene Aliases
H-FABP, O-FABP, cb889, fabp11
Chromosome Location
Chr.19: 42983620 - 42993278 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152961.3 NP_694493.1 1 - 2 192 96
GenBank mRNA AF448057.1 - 1 - 2 89 96
BC049060.1 - 1 - 2 192 96
CT725749.2 - 3 - 4 579 96
EH456910.1 - 1 - 2 221 96

View Full Product Details