Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stm
Entrez Gene ID
386700
Gene Name
starmaker
Gene Aliases
cb885, id:ibd5112, sb:cb148, sb:cb568, sb:cb584
Chromosome Location
Chr.19: 3137515 - 3167850 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198817.2 NP_942112.2 16 - 17 714 90
XM_005159327.4 XP_005159384.1 15 - 16 781 90
GenBank mRNA AY372242.1 - 16 - 17 729 90
BC129155.1 - 16 - 17 714 90

View Full Product Details