Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ammecr1
Entrez Gene ID
393553
Gene Name
AMMECR nuclear protein 1
Gene Aliases
zgc:66100, zgc:85865
Chromosome Location
Chr.21: 44709304 - 44731829 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200581.1 NP_956875.1 1 - 2 412 63
GenBank mRNA BC056704.1 - 1 - 2 412 63
BC067667.1 - 1 - 2 448 63

View Full Product Details