Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pou2f1b
Entrez Gene ID
323702
Gene Name
POU class 2 homeobox 1b
Gene Aliases
fc07d06, wu:fc07d06, zgc:158360
Chromosome Location
Chr.9: 34319333 - 34355548 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089329.1 NP_001082798.1 13 - 13 4753 157
XM_005167812.3 XP_005167869.1 14 - 14 4824 158
XM_005167813.3 XP_005167870.1 14 - 14 4821 158
XM_005167815.3 XP_005167872.1 14 - 14 4815 158
XM_005167816.3 XP_005167873.1 14 - 14 4812 158
XM_005167818.3 XP_005167875.1 13 - 13 4758 158
XM_009304813.2 XP_009303088.1 14 - 14 4812 158
XM_009304814.2 XP_009303089.1 14 - 14 4803 158
XM_017357650.2 XP_017213139.1 14 - 14 4978 158
XM_017357651.2 XP_017213140.1 14 - 14 4800 158
GenBank mRNA BC134808.1 - 13 - 13 4753 157

View Full Product Details