Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpm6ba
Entrez Gene ID
368225
Gene Name
glycoprotein M6Ba
Gene Aliases
zgc:136505
Chromosome Location
Chr.1: 28959723 - 29045423 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203427.2 NP_982251.1 7 - 7 2220 118
GenBank mRNA AY070265.2 - 9 - 9 2051 118

View Full Product Details