Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftr53
Entrez Gene ID
368624
Gene Name
finTRIM family, member 53
Gene Aliases
si:busm1-57i23.1, si:ch211-221b16.8, si:dz57i23.1
Chromosome Location
Chr.9: 609564 - 611951 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037706.1 1 - 1 544 143
GenBank mRNA BC115239.1 - 1 - 1 544 143

View Full Product Details