Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ube2e2
Entrez Gene ID
445100
Gene Name
ubiquitin-conjugating enzyme E2E 2
Gene Aliases
wu:fb38h07, zgc:92467
Chromosome Location
Chr.16: 50007000 - 50076303 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003494.1 NP_001003494.1 6 - 6 843 124
GenBank mRNA BC076483.1 - 6 - 6 843 124
BC150448.1 - 6 - 6 911 124

View Full Product Details