Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
insm1b
Entrez Gene ID
323882
Gene Name
insulinoma-associated 1b
Gene Aliases
INSM-1, Insm1, fc13a09, wu:fc13a09, zINSM-1, zgc:63866
Chromosome Location
Chr.17: 41463942 - 41466140 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199658.1 NP_955952.1 1 - 1 2046 134
GenBank mRNA AF411143.1 - 1 - 1 968 134
BC053119.1 - 1 - 1 2046 134

View Full Product Details