Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rgs8
Entrez Gene ID
436804
Gene Name
regulator of G protein signaling 8
Gene Aliases
zgc:92913
Chromosome Location
Chr.6: 205945 - 231665 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002531.1 NP_001002531.1 4 - 5 377 109
GenBank mRNA BC076365.1 - 4 - 5 377 109
BC165890.1 - 2 - 3 199 109

View Full Product Details