Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs2
Entrez Gene ID
436935
Gene Name
regulator of G protein signaling 2
Gene Aliases
fa96c08, wu:fa96c08, wu:fj08f11, zgc:91965
Chromosome Location
Chr.22: 24246213 - 24248422 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002662.2 NP_001002662.2 3 - 4 283 74
GenBank mRNA BC076456.1 - 3 - 4 269 74

View Full Product Details