Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sh3bgr
Entrez Gene ID
445130
Gene Name
SH3 domain binding glutamate-rich protein
Gene Aliases
zgc:100896
Chromosome Location
Chr.15: 231937 - 251586 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003524.2 NP_001003524.1 4 - 5 533 70
NM_001362477.1 NP_001349406.1 4 - 5 533 70
NM_001362478.1 NP_001349407.1 4 - 5 533 70
XM_009291457.3 XP_009289732.1 4 - 5 528 70
XM_021481383.1 XP_021337058.1 4 - 5 528 70
XM_021481384.1 XP_021337059.1 4 - 5 528 70
GenBank mRNA BC077137.1 - 4 - 5 506 70

View Full Product Details