Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sh3bgr
Entrez Gene ID
445130
Gene Name
SH3 domain binding glutamate-rich protein
Gene Aliases
zgc:100896
Chromosome Location
Chr.15: 231937 - 251586 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003524.2 NP_001003524.1 6 - 7 575 76
XM_005169682.4 XP_005169739.1 7 - 8 744 76
XM_005169686.4 XP_005169743.1 6 - 7 629 76
XM_005169688.4 XP_005169745.1 5 - 6 482 76
XM_009291452.3 XP_009289727.1 7 - 8 744 76
GenBank mRNA BC077137.1 - 6 - 7 548 76

View Full Product Details