Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glrbb
Entrez Gene ID
445193
Gene Name
glycine receptor, beta b
Gene Aliases
glrb2, zgc:101041
Chromosome Location
Chr.14: 239938 - 280269 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003587.1 NP_001003587.1 7 - 8 1051 115
XM_021481003.1 XP_021336678.1 7 - 8 1064 115
GenBank mRNA AB195560.1 - 6 - 7 751 115
BC078269.1 - 7 - 8 1051 115

View Full Product Details