Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc25a43
Entrez Gene ID
796731
Gene Name
solute carrier family 25 member 43
Gene Aliases
si:bz46j2.1, si:rp71-46j2.1, wu:fc65c10, zgc:101590
Chromosome Location
Chr.14: 32884731 - 32893756 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004497.2 NP_001004497.2 2 - 3 601 121
GenBank mRNA BC085377.1 - 2 - 3 601 121
BC154347.1 - 2 - 3 630 121

View Full Product Details