Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arrdc1a
Entrez Gene ID
100006005
Gene Name
arrestin domain containing 1a
Gene Aliases
arrdc1, sb:cb786, si:rp71-1o1.5, wu:fc25g09, wu:fi04h05, zgc:92248
Chromosome Location
Chr.5: 29516843 - 29531948 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004561.1 NP_001004561.1 6 - 7 877 91
XM_005171783.4 XP_005171840.1 6 - 7 862 91
GenBank mRNA BC081631.1 - 6 - 7 831 91
BC096962.1 - 6 - 7 856 91
BC152620.1 - 6 - 7 846 91
BC165815.1 - 6 - 7 825 91

View Full Product Details