Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gins1
Entrez Gene ID
450061
Gene Name
GINS complex subunit 1 (Psf1 homolog)
Gene Aliases
zgc:101672
Chromosome Location
Chr.13: 7164345 - 7170352 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006081.1 NP_001006081.1 1 - 2 178 105
GenBank mRNA BC083239.1 - 1 - 2 178 105

View Full Product Details