Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpkow
Entrez Gene ID
368735
Gene Name
G patch domain and KOW motifs
Gene Aliases
mp:zf637-1-001655, si:dz150f13.5, t54l, wu:fb38d03, zgc:152714
Chromosome Location
Chr.8: 36591772 - 36618080 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007207.1 NP_001007208.1 10 - 11 1409 92

View Full Product Details