Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem86a
Entrez Gene ID
541385
Gene Name
transmembrane protein 86A
Gene Aliases
im:7137034, zgc:113571
Chromosome Location
Chr.25: 24250248 - 24262445 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013512.2 NP_001013530.1 2 - 3 453 70
GenBank mRNA BC090488.1 - 2 - 3 460 70

View Full Product Details