Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sostdc1a
Entrez Gene ID
550261
Gene Name
sclerostin domain containing 1a
Gene Aliases
zgc:110293
Chromosome Location
Chr.15: 34562194 - 34567365 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017598.2 NP_001017598.2 1 - 2 398 78
GenBank mRNA BC092855.1 - 1 - 2 261 78
BC152132.1 - 1 - 2 267 78
BC164672.1 - 1 - 2 233 78
EH592789.1 - 1 - 2 287 78
EH605481.1 - 1 - 2 206 78

View Full Product Details