Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
shisa4
Entrez Gene ID
550567
Gene Name
shisa family member 4
Gene Aliases
zgc:110683
Chromosome Location
Chr.11: 58760 - 65378 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017869.1 NP_001017869.1 2 - 3 415 64
GenBank mRNA BC092979.1 - 2 - 3 415 64

View Full Product Details