Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
myl10
Entrez Gene ID
550569
Gene Name
myosin, light chain 10, regulatory
Gene Aliases
id:ibd2025, mlc10, zgc:110679
Chromosome Location
Chr.10: 33171862 - 33179915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017871.1 NP_001017871.1 3 - 4 217 105
GenBank mRNA BC092977.1 - 3 - 4 217 105

View Full Product Details