Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankrd29
Entrez Gene ID
553798
Gene Name
ankyrin repeat domain 29
Gene Aliases
si:dkey-121a11.5, zgc:111996
Chromosome Location
Chr.22: 16418695 - 16431136 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020760.1 NP_001018596.1 4 - 5 588 55
XM_005161708.4 XP_005161765.1 4 - 5 449 55
GenBank mRNA BC095639.1 - 4 - 5 588 55

View Full Product Details