Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
b9d1
Entrez Gene ID
554150
Gene Name
B9 protein domain 1
Gene Aliases
im:6905750, im:7069491, wu:fc09g07, zgc:110733
Chromosome Location
Chr.3: 39691530 - 39696058 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024373.1 NP_001019544.1 2 - 3 260 133
GenBank mRNA BC095392.1 - 2 - 3 260 133

View Full Product Details