Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nitr11b
Entrez Gene ID
558014
Gene Name
novel immune-type receptor 11b
Gene Aliases
nitr11b_1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025497.1 NP_001020668.1 2 - 3 392 139

View Full Product Details