Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ublcp1
Entrez Gene ID
792145
Gene Name
ubiquitin-like domain containing CTD phosphatase 1
Gene Aliases
fb33g09, wu:fb33g09, zgc:86634
Chromosome Location
Chr.21: 35304974 - 35325541 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002319.2 NP_001002319.1 11 - 12 1084 139
NM_001199375.1 NP_001186304.1 10 - 11 1000 139
XM_021468921.1 XP_021324596.1 11 - 12 1239 139

Gene Symbol
LOC103910001
Entrez Gene ID
103910001
Gene Name
ubiquitin-like domain-containing CTD phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 4282 - 16347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009298383.3 XP_009296658.2 9 - 10 897 140

View Full Product Details