Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nubpl
Entrez Gene ID
436715
Gene Name
nucleotide binding protein-like
Gene Aliases
zgc:92633
Chromosome Location
Chr.17: 31719046 - 31726868 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002442.1 NP_001002442.1 10 - 11 1128 67
GenBank mRNA BC076121.1 - 10 - 11 1128 67
BC153444.1 - 10 - 11 1149 67

View Full Product Details