Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
txnl4b
Entrez Gene ID
641329
Gene Name
thioredoxin-like 4B
Gene Aliases
CH73-41E3.4, zgc:123253
Chromosome Location
Chr.1: 513982 - 522317 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037123.3 NP_001032200.1 3 - 4 550 141
GenBank mRNA BC107846.1 - 3 - 4 504 141
EH498384.1 - 3 - 4 561 141

View Full Product Details