Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnd2
Entrez Gene ID
751645
Gene Name
Rho family GTPase 2
Gene Aliases
Rho, arhn, si:dz163l24.1, zgc:153204
Chromosome Location
Chr.3: 319581 - 368526 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045364.1 NP_001038829.1 4 - 5 552 87
GenBank mRNA BC122188.1 - 4 - 5 552 87
BC154818.1 - 3 - 4 586 87
BC165068.1 - 4 - 5 469 87

View Full Product Details