Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snx22
Entrez Gene ID
561624
Gene Name
sorting nexin 22
Gene Aliases
im:6905586, zgc:153718
Chromosome Location
Chr.25: 5992789 - 6011049 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045374.2 NP_001038839.2 3 - 4 454 141
GenBank mRNA BC122395.1 - 3 - 4 423 141

View Full Product Details