Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fut11
Entrez Gene ID
568888
Gene Name
fucosyltransferase 11 (alpha (1,3) fucosyltransferase)
Gene Aliases
zgc:153537
Chromosome Location
Chr.14: 14608100 - 14622984 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077174.1 NP_001070642.1 2 - 3 792 120
GenBank mRNA BC124380.1 - 2 - 3 792 120

View Full Product Details