Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ldlrap1b
Entrez Gene ID
791153
Gene Name
low density lipoprotein receptor adaptor protein 1b
Gene Aliases
zgc:158745
Chromosome Location
Chr.13: 45582367 - 45647639 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080635.1 NP_001074104.1 1 - 2 276 60
XM_021480671.1 XP_021336346.1 1 - 2 300 60
GenBank mRNA BC129425.1 - 1 - 2 276 60
BC164254.1 - 1 - 2 124 60

View Full Product Details