Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dachd
Entrez Gene ID
791191
Gene Name
dachshund d
Gene Aliases
si:ch211-176j8.3, zgc:158869
Chromosome Location
Chr.9: 30890549 - 31065163 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080673.1 NP_001074142.1 5 - 6 1346 138
XM_005167749.4 XP_005167806.1 5 - 6 1341 138
XM_005167750.4 XP_005167807.1 5 - 6 1341 138
GenBank mRNA BC129504.1 - 5 - 6 1346 138

View Full Product Details