Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grhl2b
Entrez Gene ID
100034503
Gene Name
grainyhead-like transcription factor 2b
Gene Aliases
grhl2, si:dkey-21k18.2
Chromosome Location
Chr.19: 12234751 - 12290999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083072.1 NP_001076541.1 3 - 4 615 131
XM_005159580.4 XP_005159637.1 4 - 5 912 131

View Full Product Details