The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083099.1 | NP_001076568.1 | 5 - 6 | 1561 | 79 |
XM_009293696.2 | XP_009291971.1 | 5 - 6 | 1698 | 79 |